missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Total Nitrogen Test Kits

Product Code. 11754725
Click to view available options
Range:
0.5 to 15 mg/L
0.5 to 15 mg/L N Total
10 to 150 mg/L
Unit Size:
0.15kg
0.25kg
Each
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11754725

Brand: WTW™ 250494

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Obtain precision results using the CombiCheck Total Nitrogen NTotal cell ready-made test kit for the determination of total nitrogen in waste water, drinking water, and surface water.

  • Variety of instrumentation for testing nitrogen in water
  • For waste water, drinking water, surface water
  • Nitrospectral Method
  • Photometric Test Methods
  • CombiCheck Total Nitrogen NTotal cell test kit
  • Safe determinations of COD and NTotal in water
TRUSTED_SUSTAINABILITY

Specifications

Analytes Chromosulfuric acid oxidation
For Use With (Application) For waste water, drinking water, surface water
For Use With (Equipment) For PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers
Includes Certificates
No. of Tests 48
Range 10 to 150 mg/L
Sufficient For 25 Tests
Test Method Photometric
Type CombiCheck COD Chemical oxygen demand O2 cell test kit
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nitrogen (total) Cell Test Method: photometric, DMP 10 - 150 mg/l N Spectroquant
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Oxidising solid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, phosphoric acid; Potassium persulfate, Potassium carbonate; sodium hydroxide; 2-Prop
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.