Learn More
Bellingham and Stanley™ Touch Screen Refractometer
Peltier temperature controlled refractometer with resolution touch display
£9370.40 - £10038.20
Specifications
Type | Digital Refractometer |
---|---|
ScaleRange | 0 to 100 Brix |
Includes | Refractometer, RFID Tags (2), Instruction Manual, and Certificate of Conformity |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
16614912
|
Bellingham and Stanley™
19-30 |
Resolution: 0.01 Brix |
£9370.40
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
15807205
|
Bellingham and Stanley™
19-40 |
Resolution: 0.01/0.001 Brix |
£10038.20
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
Description
Touch Screen Refractometer with Peltier temperature control features the latest integral 7 in. full color touchscreen, high definition display, and USB/LAN connectivity. The refractometer is supplied with two RFID tags, instruction manual, and certificate of conformity. Perfect for measurement and control in demanding food, beverage, chemical and industrial applications.
- Scales
- RI 1.32 - 1.58
- Brix 0 - 100
- Resolution:
- RI 0.00001 (RFM330-T)/ 0.000001 (RFM340-T)
- Brix 0.01
Specifications
Digital Refractometer | |
0 to 100 Brix | |
Refractometer, RFID Tags (2), Instruction Manual, and Certificate of Conformity |