missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ TouchNTuff™ 73-300 Series Neoprene Gloves
GREENER_CHOICEBIOPHARMA

Product Code. 15249189
Click to view available options
Size:
7
9
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides unprecedented sensitivity in double-gloving and dry applications due to the 0.13mm (5 mil) polymer thickness. Ansell TouchNTuff™ 73-300 Series Neoprene Gloves offer splash resistance against a broad range of chemicals.

  • Ergonomically designed, hand-specific formers means superior fit and comfort, less hand fatigue and stress.
  • Made from 100% polychloroprene
  • Hypoallergenic - Contains no natural rubber latex or accelerators used in manufacturing reducing the risk of Type I and IV skin allergies
  • Ansell SureFit Technology eliminates cuff roll-down for even better protection
  • Polyurethane inner coating enables easy donning with dry or wet hands
  • Design and polymer thickness facilitate ease and comfort of double gloving
  • The cream color reveals breaches when used with gloves of another color
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 374:2003
Classification Category III
Coated Area Inner lining
Coating Material Polyurethane
Color Cream
Disposable Single-use
Exterior Finish Smooth
Latex Free Yes
Material Neoprene, Polychloroprene
Product Type Chemical Resistant Gloves
Size 7
Sterility Non-sterile
Thickness (English) 5 mil
Thickness (Metric) 0.13 mm
Glove Cuff Style Beaded/Rolled
Quantity 200 pairs/carton
Series 73-300
Length (Metric) 305 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'