Learn More
Ansell TouchNTuff™ 83-500 Sterile Polyisoprene Gloves
GREENER_CHOICEBIOPHARMA
Product Code. 15156563
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
Sterile polyisoprene gloves are ergonomically-designed to enhance fit and comfort, and reduce hand fatigue and stress.
  • Powder-free, chlorinated glove interior
  • Rolled bead cuff style
  • Hand specific
  • Smooth finish
  • Polyisoprene
  • White
TRUSTED_SUSTAINABILITY
Specifications
Cleanroom Class Class 100 (ISO 5)
Exterior Finish No Texture
Product Type Cleanroom Gloves
Size 6
Sterility Sterile
Thickness (Metric) 0.21 mm
Glove Cuff Style Beaded/Rolled
Model 835006
Quantity 200/Pk.
Resistant To Splash resistance
Series 83-500