missing translation for 'onlineSavingsMsg'
Learn More

Ansell TouchNTuff™ 93-163 Series Blue Disposable Nitrile Gloves

Product Code. 15552505
Click to view available options
Size:
10.5/11
6.5/7
7.5/8
8.5/9
9.5/10

Proven splash resistance against hazardous chemicals. Ansell TouchNTuff™ 93-163 Series Blue Disposable Nitrile Gloves are flexible and comfortable offering better dexterity.

  • Single use gloves
  • Nitrile formulation for low hand fatigue
  • Easy donning
  • Textured finish, light and resistant
  • Good resistance to puncture
  • Flexible and comfortable offering excellent dexterity
  • Beaded cuff for an easy donning and offering good tear resistance
  • Ambidextrous

Applications:

  • Chemical handling
  • Glass manufacturing
  • Handling of cytostatics
  • Intricate parts handling
  • Laboratory analysis
  • Light assembly of oil-coated pieces
  • Light assembly tasks
  • Paint and spray shops
  • Pharmaceuticals
  • Printing industry
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 374:2003
Color Blue
Disposable Single-use
Exterior Finish Fully Textured
Material Nitrile
Powdered Powder Free
Product Type Nitrile Gloves
Size 8.5/9
Thickness (Metric) 0.17 mm
Glove Cuff Style Beaded/Rolled
Packaging Type 50 gloves in a box, 10 boxes in a carton
Series 93-163
Length (Metric) 355 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'