missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ TouchNTuff™ 93-300 Series Green Nitrile Gloves
Complies with ASTM D 3577, Type II. Ansell™ TouchNTuff™ 93-300 Series Green Nitrile Gloves are ideal for use in electronics and semiconductor manufacturing, and laboratory testing.
£217.80 - £480.00
Specifications
Back Material | Nitrile |
---|---|
Certifications/Compliance | Standards CATEGORY III EN 374, EN 421, ASTM D 3577, Type II |
Cuff Material | Nitrile |
Coating Material | Nitrile |
Latex Free | Yes |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15254698
|
Ansell™
93300080 |
Medium |
£480.00
Pack of 1000 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15284698
|
Ansell™
93300100 |
X-Large |
£480.00
Pack of 1000 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
15274698
|
Ansell™
93300090 |
Large |
£480.00
Pack of 1000 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
15366195
|
Ansell™
93300070 |
Small |
£480.00
Pack of 1000 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
15201588
|
Ansell™
93-300_9.5-10 |
9.5 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Delivers the highest protection for a wide range of chemicals than any other single use nitrile glove
- Provides the wearer with a high level of comfort and increased protection in the forearm (longer cuff)
- Manufactured from nitrile, offering three times the puncture resistance of standard latex gloves
- Made without natural rubber latex proteins, the glove may be worn with confidence by latex sensitive (Type l) wearers
- Suitable for ISO Class 5 or higher clean room environments
Specifications
Nitrile | |
Nitrile | |
Yes | |
0.12 mm | |
For blending of liquids, filtration processes, vaccines injectables operations etc. |
Standards CATEGORY III EN 374, EN 421, ASTM D 3577, Type II | |
Nitrile | |
Gloves | |
Textured fingertips | |
Splash and high puncture resistance |