missing translation for 'onlineSavingsMsg'
Learn More

Excel Healthcare Ltd™ Tower 200 Water Distiller

Produces distilled water efficiently

Brand:  Excel Healthcare Ltd™ 287

Encompass

Product Code. 16310132

  • POA / Each

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Distilled Water Is Purified Water That Has Been Boiled Into Steam And Condensed Back Into Liquid In A Separate Container
  • Impurities In The Original Water That Do Not Boil Below The Boiling Point Of Water Remain In The Original Container
  • Distilled Water Is One Type Of Purified Water
Specifications

Specifications

4 L
565 W/ Cooling Fan 15 W
Stainless Steel Tank and Lid, Plastic Shell
Water Distiller Output 16 L/day
230 x 280 x 445 mm
Power Cord (UK), 4 L Collector Bottle/Jug, 250 g Residue Cleaner
230/240 VAC
Water Distiller, Tower 200
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.