missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toxaphene Standard, High Level, SPEX CertiPrep™
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Toxaphene | 8001-35-2 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
CAS | 8001-35-2, 110-54-3 |
Packaging | 2mL amber prescored glass ampule |
CAS Min % | 0.2, 99.8 |
Quantity | 1 mL |
CAS Max % | 0.2, 99.8 |
Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
Methods | For US EPA Method 625 |
Solvent or Matrix | Hexane |
Chemical Name or Material | Toxaphene Standard (High Level), USEPA Method 625 |
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'