missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toxicity Characteristic Leachate Procedure Standard 1, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£175.00 - £261.00
Specifications
Chemical Name or Material | TCLP Standard 1 without Mercury |
---|---|
Name Note | Without Mercury |
Packaging | Triple-leached LDPE bottle |
CAS | 7440-39-3, 7440-22-4, 7440-38-2, 7697-37-2, 7732-18-5 |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15266326
|
SPEX Certiprep
TCLP-100N |
125 mL | Triple-leached LDPE bottle |
£175.00
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15276326
|
SPEX Certiprep
TCLP-500N |
500 mL | Triple-leached LDPE bottle |
£261.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ba | 500μg/mL |
Ag | 25μg/mL |
As | 25μg/mL |
Cr | 25μg/mL |
Pb | 25μg/mL |
Cd | 5μg/mL |
Se | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
TCLP Standard 1 without Mercury | |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Assurance | |
2% HNO3 |
Without Mercury | |
7440-39-3, 7440-22-4, 7440-38-2, 7697-37-2, 7732-18-5 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Ba( 500mg/L),Ag(25mg/L),As(25mg/L) |
Safety and Handling
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only