Learn More
Lonza™ ToxiLight™ Non-Destructive Cytotoxicity BioAssay Kit, 500 test
Designed to measure toxicity in mammalian cells and cell lines in culture.
Brand: Lonza™ LT07-217
This item is not returnable.
View return policy
Description
- Safe - No hazardous chemicals are required.
- Flexible - Cell supernatants can be frozen with no loss of AK activity, allowing for long-term kinetic studies.
- Fast - Readable signal is detectable 5 minutes after the addition of the ToxiLight™ BioAssay reagent.
- Simple - Assay requires the simple addition of a single reagent, which can be added directly to wells in which cells are growing, or to a small sample of aspirated culture supernatant.
- Non-destructive - ToxiLight™ assay detects cellular AK present in cell culture supernatants, eliminating the need to lyse cells to perform the assay, and allowing multiple tests to be performed on the same sample.
- Highly sensitive - The ToxiLight™ BioAssay Kit exploits the cyclic nature of the AK reaction whereby a small amount of the AK enzyme can generate high concentrations of ATP. This enables the detection of very subtle changes in cytotoxicity and reduces false negatives.
Specifications
Non-destructive cytotoxicity | |
100 tests/ 5 plates | |
LT27-060 AK detection reagent, LyophiLized, 5 x 10 mL vials, LT27-066 AK assay buffer 1 x 50 mL bottLe |
LT27-060 AK detection reagent, LT27-066 AK assay buffer | |
Cytotoxicity assay |