missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Toyocamycin
Adenosine analog; antifungal antibiotic
£71.00 - £236.00
Specifications
Purity | >98% |
---|---|
Product Type | Toyocamycin |
Molecular Weight (g/mol) | 291.26 |
Inhibitors | Toyocamycin |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18764719
|
Tocris Bioscience™
5270/10 |
10 mg |
£71.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18720268
|
Tocris Bioscience™
5270/50 |
50 mg |
£236.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adenosine analog; antifungal antibiotic. Inhibits RNA self-cleavage in HEK79 cells and PI 3-kinase activity in A431 epidermoid carcinoma cell membrane fractions. Also inhibits thapsigargin-induced XBP1-luciferase activation and induces apoptosis in multiple myeloma cell lines.Specifications
>98% | |
Toyocamycin | |
291.26 | |
Toyocamycin |
For Research Use Only