missing translation for 'onlineSavingsMsg'
Learn More

TPI1 Mouse anti-Human, Unconjugated, Clone: 2E7E5, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen TPI1
Clone 2E7E5
Concentration 1000 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16880186
View Documents
Proteintech
67515-1-IG-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16870186
View Documents
Proteintech
67515-1-IG-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The TPI1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TPI1. This antibody recognizes human, mouse, rat antigen. The TPI1 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

TPI1
1000 μg/mL
Monoclonal
Mouse
7167
Primary
2E7E5
Western Blot, ELISA
Unconjugated
Human, Rat, Mouse
IgG1
Videos
SDS
Documents

Documents

Product Certifications