missing translation for 'onlineSavingsMsg'
Learn More
TPL 4 Model Buffer Solution pH 4, WTW™ Product Code.: 11765154
compliance-icons compliance-icons compliance-icons

TPL 4 Model Buffer Solution pH 4, WTW™

Product Code. 11765154
250 mL
Click to view available options
Quantity:
250 mL
This item is not returnable. View return policy

Product Code. 11765154

Brand: WTW 108800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These modern technical buffering solutions that are traceable to NIST (National Institute of Standards and Technology, are available in the practical 250 ml bottle including dosing device TPL 4 and live up to 3 years (unopened) through thrifty use.

  • pH buffers by WTW – precise and traceable to PTB/NIST in bulk size of 250 ml with built-in dosing vessel standard buffer
  • Live up to 3 years (unopened) through thrifty use
  • Easiest handling by means of dosing device
  • Technical buffering solutions for pH 4.01 that is available in the practical 250 ml bottle including dosing device TPL 4
  • This modern technical buffering solution is traceable to NIST (National Institute of Standards and Technology, USA)
  • Their accuracy is ± 0,03 pH
TRUSTED_SUSTAINABILITY

Specifications

pH 4
Physical Form Liquid
Quantity 250 mL
Packaging Bottle
Traceability to NIST Yes
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc acetate dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contains : Zinc di(acetate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TPL 4 Model Buffer Solution pH 4, WTW™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.