Learn More
TR-FRET Coregulator Buffer A
TR-FRET Coregulator Buffer A
Brand:  Invitrogen PV4384
Code : 50
Additional Details : Weight : 0.50000kg
Product Code. 10562704
  • £130.00 / Each
Estimated Shipment: 05-08-2024
to see stock.
For Research Use Only. All usage must comply with product instructions.
Description
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Specifications
Specifications
TR-FRET Coregulator Buffer A, 25mL, Liquid, 7.5 pH, 20 to 30°C Storage, Room Temperature or on Wet or Dry Ice Shipping Conditions, Water Soluble, For use with Pharma and Biopharma, Target and Lead Identification and Validation
25 mL