Learn More
Thermo Scientific™ TR-FRET Coregulator Buffer D
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Brand: Thermo Scientific™ PV4420
This item is not returnable.
View return policy
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Specifications
TR-FRET Coregulator Buffer D, 25mL, Liquid, 7.5 pH, 4°C Storage, Room Temperature or on Wet or Dry Ice Shipping Conditions, Water Soluble, For use with Protein Assays and Analysis, High Throughput-Compatible, Enzyme Assay | |
Liquid | |
Water | |
7.5 | |
TR-FRET Coregulator Buffer D may be shipped frozen or at room temperature. Store at 4°C. |
Pharma and biopharma, target and lead identification and validation, Lanthascreen™ nuclear receptor cofactor interaction assays | |
25mL, 25 mL | |
Nuclear Receptor Buffer | |
LanthaScreen™ Nuclear Receptor Cofactor Interaction Assays | |
Approved for shipment at Room Temperature or on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.