Learn More
TR-FRET Coregulator Buffer E
TR-FRET Coregulator Buffer E
Brand: Invitrogen PV4540
Code : 50
Additional Details : Weight : 0.10500kg
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.Specifications
TR-FRET Coregulator Buffer | |
25 mL | |
Liquid |
TR-FRET Coregulator Buffer E may be shipped frozen or at room temperature. It should be stored at room temperature (20°C to 30°C). | |
Approved for shipment at Room Temperature or on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.