Learn More
TR-FRET Coregulator Buffer J
TR-FRET Coregulator Buffer J
Brand:  Invitrogen PV4682
Product Code. 10223848
  • £127.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TR-FRET Coregulator Buffer J may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C).
Liquid
25 mL
Approved for shipment at Room Temperature or on Wet or Dry Ice