Learn More
TR-FRET Dilution Buffer
TR-FRET Dilution Buffer
Brand:  Invitrogen PV3574
Product Code. 10398323
  • £314.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
TR-FRET Dilution Buffer is an essential component for kinase assays using LanthaScreen™ technology. The TR-FRET Dilution Buffer is used to dilute LanthaScreen™ antibodies for screening applications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dilution Buffer
TR-FRET Dilution Buffer may be shipped frozen or at room temperature. Store at room temperature (20°C to 30°C).
Approved for shipment at Room Temperature or on Wet or Dry Ice
ABL1 G250E, CSF1R, CSK, EPHA5, KIT, PDGFRA T674I, PDGFRA V561D, PTK6, RET Y791F, SYK, TYRO3, ZAP70, MAP4K5, MST4, PRKCQ, PRKX, ROCK2, IKBKE, PAK4, EIF2AK1, MAP3K2, LRRK2 D1994A, RAF1 (cRAF) Y340D Y341D, FGFR1, FGFR2, PRKCD, MAPK13, MAPK8, MAP3K10, MAP3K9, JAK1, JAK2 JH1 JH2, AMPK (A2/B1/G1), CAMK1, CAMK2B, CHEK1, MAPKAPK2, PASK, PIM2, PRKCN, PRKD2, PRKG2, RPS6KA5, BLK, EGFR, EPHB2, FGFR4, IGF1R, LYNA, MATK, MERTK, NTRK3, RET V804L, ROS1, TEK, TTK, NEK2, PRKCG, SGK, CHUK, IKBKB, TAOK3, JAK2 JH1 JH2 V617F, TYK2, DNAPK, FLT1, MUSK, SRCN1, TEC, CAMK2D, MAP3K7/MAP3K7IP1, LRRK2 R1441C, ABL1 E255K, BMX, EPHA4, FYN, PTK2, PTK2B, RET, MAP4K2, MINK1, SGK2, GRK6, MAPK9, JAK3, LRRK2, AMPK (A1/B1/G1), AURKA, CAMK1D, CAMK4, CHEK2, MAPKAPK3, PIM1, PKN1, PRKD1, PRKG1, RPS6KA4, STK22D, ABL1 T315I, EPHA3, EPHB4, FLT3, FLT4, INSR, LCK, YES1, EGFR L858R, ERBB4, KDR, MET, TNK2, PRKCA, PRKCI, CAMK2A, PAK1, RPS6KA3, MAPK14, FRAP1, EIF2AK3, MAP3K3, ABL1, ABL2, AXL, FGFR3, FGFR3 K650E, FGR, HCK, KIT V654A, AURKB, TAOK2, MAP3K11, NLK, MAPK10, EPHA1, EPHA8, EPHB3, INSRR, LYNB, MST1R, NTRK1, PDGFRA D842V, SRMS, MAP4K4, AKT2, mTOR, LRRK2 G2019S, EGFR L861Q, EPHA2, FER, LTK, PDGFRA, PDGFRB, MARK4, RPS6KB1, EGFR T790M, EPHA7, EPHB1, MET M1250T, DYRK4, PHKG1, PRKCZ, ROCK1, RPS6KA1, TBK1, MAPK1, MAPK11, BRAF V599E, JAK2, FRK, ITK, PRKCB1, STK24, AKT3, BRAF, LRRK2 I2020T, FLT3 D835Y, SRC, GRK5, ABL1 Y253F, BTK, DDR2, FES, KIT T670I, TXK, PRKCB2, RPS6KA2, SGKL, STK25, AKT1, PAK7, PRKACA, MAPK12, MAPK3, MAP3K8, ERBB2, NTRK2, PRKCE, PRKCH, EIF2AK2, MAP3K5
100 mL