Learn More
TR40 Concentrate
TR40 Concentrate
Brand:  Invitrogen K1031
Code : 50
Additional Details : Weight : 3.20000kg
Product Code. 10696645
  • £2520.00 / Each
Estimated Shipment: 16-09-2024
to see stock.
For Research Use Only. All usage must comply with product instructions.
Description
Description
TR40 Concentrate is the suppression dye component of Solution C in the Loading Kits for CCF2-AM and CCF4-AM.
Specifications
Specifications
CCF2-AM and CCF4-AM
Store at room temperature.
GeneBLAzer™
1 L
Room Temperature