missing translation for 'onlineSavingsMsg'
Learn More

Trace Metals 1 without Mercury For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep WP-15-500N

Code : NEW

Additional Details : Weight : 0.50000kg Transport : UN number : 3264 Chem class : 8 Pack group : III

 View more versions of this product

Product Code. 15206346

  • £424.00 / 500mL
Estimated Shipment: 02-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al500μg/mL
V250μg/mL
As100μg/mL
Be100μg/mL
Co100μg/mL
Cr100μg/mL
Cu100μg/mL
Fe100μg/mL
Mn100μg/mL
Ni100μg/mL
Pb100μg/mL
Zn100μg/mL
Cd25μg/mL
Se25μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Trace Metals I without Mercury
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Environmental
5% HNO3
Groundwater and Wastewater Pollution Control Check Standard
500 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Various
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product intended for laboratory use only