missing translation for 'onlineSavingsMsg'
Learn More

Trace Metals 3 For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep MN-6


 View more versions of this product

Product Code. 11425938

  • £206.00 / 125mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
This item is not returnable. View return policy

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ba500μg/mL
Ca500μg/mL
Mo500μg/mL
Na500μg/mL
K100μg/mL
Mg100μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Trace Metals Standard 3
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Environmental
2% HNO3
Groundwater and Wastewater Pollution Control Check Standard
125 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Components in 2% HNO3:
Ba    •500μg/mL
Ca    •500μg/mL
Mg    •100μg/mL
Mo    •500μg/mL
K    •100μg/mL
Na    •500μg/mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Trace Metals III

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

Supplemental information
  • MIXTURE LIST-Contains : nitric acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Product intended for laboratory use only