Learn More
PureAire Monitoring Systems, Inc. Trace Oxygen Analyser, 0-1000ppm
For use in a vacuum environment, 3D printer, nitrogen generator, or semiconductor tool using nitrogen, argon, or cryogenics. The robust zirconium sensor is connected via the KF25 flange. Sensor is connected to the analyser using a 3m cable for easy connection.
Brand: PureAire Monitoring Systems, Inc.
Additional Details : Weight : 0.91260kg
Description
- Accurate ppm Zirconium Sensor
- Two-Alarm setpoints
- Built-in audible alarm 90 db
- Digital display, and 4-20mA analog output
- Wall mounting brackets
Why choose PureAire?
- As an industry leader in oxygen analyser and gas detection, PureAire is dedicated to the safety of our clients, and the customers they serve, by manufacturing monitors that are long-lasting, innovative, and reliable.
What is an O2 Analyser?
- The Air Check Trace O2 Analyser is designed for continuous monitoring low oxygen levels need from zero to 1,000 parts per million (ppm)
What gases can an O2 monitor be used for?
- PureAire’s oxygen analysers can be used for nitrogen, and argon.
Certifications
CE Marked
Services
2 years
Specifications
3 in. | |
5.125 x 3.15 x 3 in. | |
80 mm | |
-40°F to +134°F | |
24V | |
1.1 lb. | |
5.125 in. |
76 mm | |
3.15 in. | |
0-95% RH | |
-40 to +55°C | |
2 Year after ETD | |
0.5 kg | |
130 mm |