Learn More
Thermo Scientific™ TRACE™ TR-1 GC Columns
Use this non-polar GC column for method development and analysis of chlorinated and nitroaromatic compounds and environmental samples.
£327.63 - £1140.68
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
10668805
|
Thermo Scientific™
260A298P |
30 m | 0.53 mm | 1 μm | N/A | ||||
10279304
|
Thermo Scientific™
260A155P |
60 m | 0.32 mm | 0.25 μm | N/A | ||||
10374363
|
Thermo Scientific™
260A395P |
30 m | 0.32 mm | 3 μm |
£597.98
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||
10708043
|
Thermo Scientific™
260A297P |
30 m | 0.32 mm | 1 μm |
£597.98
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||
10600174
|
Thermo Scientific™
260A143P |
30 m | 0.32 mm | 0.25 μm |
£577.88
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
10070073
|
Thermo Scientific™
260A223P |
30 m | 0.25 mm | 0.5 μm |
£557.78
Each |
In Stock
Log in to see stock. |
|||
10311545
|
Thermo Scientific™
260A470P |
30 m | 0.53 mm | 5 μm |
£623.10
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
10269464
|
Thermo Scientific™
260A309P |
60 m | 0.32 mm | 1 μm | N/A | ||||
10496873
|
Thermo Scientific™
260A308P |
60 m | 0.25 mm | 1 μm |
£949.73
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||
10585225
|
Thermo Scientific™
260A224P |
30 m | 0.32 mm | 0.5 μm | N/A | ||||
10475533
|
Thermo Scientific™
260A130P |
15 m | 0.25 mm | 0.25 μm |
£327.63
Each |
In Stock
Log in to see stock. |
|||
10068942
|
Thermo Scientific™
260A131P |
15 m | 0.32 mm | 0.25 μm |
£347.73
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||
10475723
|
Thermo Scientific™
260A142P |
30 m | 0.25 mm | 0.25 μm |
£542.70
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||
10592635
|
Thermo Scientific™
260A396P |
30 m | 0.53 mm | 3 μm | N/A | ||||
10179414
|
Thermo Scientific™
260A336P |
30 m | 0.53 mm | 1.5 μm | N/A | ||||
10506005
|
Thermo Scientific™
260A241P |
100 m | 0.25 mm | 0.5 μm |
£1140.68
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
10725574
|
Thermo Scientific™
260A225P |
30 m | 0.53 mm | 0.5 μm | N/A | ||||
10671206
|
Thermo Scientific™
260A296P |
30 m | 0.25 mm | 1 μm |
£562.80
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||
10219264
|
Thermo Scientific™
260A154P |
60 m | 0.25 mm | 0.25 μm |
£949.73
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||
Description
Use the Thermo Scientific™ TRACE™ TR-1 GC column for method development and chlorinated and nitroaromatic compounds and environmental analyses. These non-polar phase, 100% dimethyl polysiloxane gas chromatography columns have a high operating temperature.
Specifications
TR-1 |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.