missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TRACE™ TR-50MS GC Columns

Choose these mid-polarity columns for GC/MS applications.

£259.00 - £707.15

Specifications

USP Type G3
Max. Temperature 360/370°C
Polarity Mid-Polar
View More Specs

Products 7
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
10350955
View Documents
Thermo Scientific™
260R047P
30 m 0.25 mm 0.1 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10576565
View Documents
Thermo Scientific™
260R130P
15 m 0.25 mm 0.25 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10618895
View Documents
Thermo Scientific™
260R142P
30 m 0.25 mm 0.25 μm
£577.88
Each
Estimated Shipment: 23-04-2025
to see stock.
Add to basket
 
10055724
View Documents
Thermo Scientific™
260R143P
30 m 0.32 mm 0.25 μm
£643.20
Each
Estimated Shipment: 10-04-2025
to see stock.
Add to basket
 
10744627
View Documents
Thermo Scientific™
260R154P
60 m 0.25 mm 0.25 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10046204
View Documents
Thermo Scientific™
260R213P
15 m 0.53 mm 0.5 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10671196
View Documents
Thermo Scientific™
260R225P
30 m 0.53 mm 0.5 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Thermo Scientific™ TRACE TR-50MS GC Columns are mid-polarity columns well-suited to GC/MS applications.

  • Mid-polarity phase, 50% Phenyl polysilphenylene-siloxane
  • Low-bleed decreases MS contamination
  • Particularly useful for applications requiring a higher temperature and more polarity than a 5% phenyl column
  • Column inertness results in minimal peak tailing and decreased breakdown of sensitive samples
  • Similar to: OV-17, SP-2250, DB-17, DB-17ms, DB-17ht, BPX50, Rtx-50, SPB-50, HP-50+, HP-17, AT50, RSL-300, PE-17, CC-17, 007-17(MPS-50), SPB-17, ZB-50

Recommended for:

  • Herbicides
  • Drugs of Abuse
  • EPA 604, 608, 8060, 8081
  • Pharmaceuticals
Specifications

Specifications

G3
360/370°C
Mid-Polar
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ TRACE™ TR-50MS GC Columns

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.