Learn More
Fisherbrand™ Traceable™ Data Logging Ethernet Thermometers Compatible with TraceableLIVE™ Cloud Service
Thermometer that allows you to stay connected to your critical environments no matter where you go.
Brand:  Fisherbrand™ 61012-17
 View more versions of this product
Product Code. 16526820
  • £264.00 / Each
Explore more special offers
Description
Description
  • Connect to your data via smartphone, tablet, or PC using TraceableLIVE cloud service
  • Get the reliability, speed, and security using Ethernet connection
  • View current temperature conditions anywhere – cloud-based interface sends email, text, and push notifications
  • No software required – web, Android™, Apple™ iOS data visibility
  • Get accurate, reliable measurements with included NIST-traceable calibration
Specifications
Specifications
Thermometer
One bottle probe with 10 ft. (3 m) long cable, holder with magnetic back, Ethernet cable, universal power adapter and plug with 5 ft. (1.5 m) long cord, three AAA alkaline batteries, and NIST-traceable calibration certificate with data
Temperature monitoring
6.985 cm
Calibrated
Plastic
-50°C to +60°C
0.544 kg