missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Traceable™ Digital Dial Thermometer
Easy to read digital dial and mercury free. Traceable™ Digital Dial Thermometers are ideal for cuvets, soil, incubators and test tubes. Wide temperature span combines the ranges of more than 90 different mercury thermometers.
£61.00
Specifications
Certifications/Compliance | ISO 17025, A2LA, NIST |
---|---|
Diameter (English) Probe | 0.19 in. |
Diameter (Metric) Probe | 4.83 mm |
For Use With (Application) | Soil, cuvettes, test tubes, food products, incubators, biomedical reagents and petroleum Hydrocarbons |
Includes | Protective Case, Silver Oxide Battery |
Product Code | Brand | Scale | Temperature Range (Metric) | Temperature Range (English) | Accuracy | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Scale | Temperature Range (Metric) | Temperature Range (English) | Accuracy | Price | Quantity & Availability | ||
15223986
|
Fisherbrand™
98769-17 |
Centigrade and Fahrenheit Scale | -50°C to +300°C | -58°F to +572°F | ±1°C (±1°F) |
£61.00
Each |
In Stock
Log in to see stock. |
||
Description
- LCD display 20 times more readable than eye-straining pointers or mercury levels
- Readings updated every 10 seconds
- Operate continuously for 1.5 years on a single replaceable battery
- Resolution, Accuracy: 0.10, ±1°C Stem length: 4.8 in. (12 cm)
- Stem diameter: 0.12 in. (3 mm)
- Dial diameter: 1.5 in. (38 mm)
- Weight: 1.01 oz. (31 g)
- Range: -58°F to +572°F and -50°C to +300°C
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
ISO 17025, A2LA, NIST | |
4.83 mm | |
Protective Case, Silver Oxide Battery | |
12 cm | |
0.1° | |
31 g | |
572°F | |
-58°F | |
-58°F to +572°F | |
Silver Oxide |
0.19 in. | |
Soil, cuvettes, test tubes, food products, incubators, biomedical reagents and petroleum Hydrocarbons | |
4.8 in. | |
Plastic | |
1 oz. | |
Thermometer | |
300°C | |
-50°C | |
-50°C to +300°C | |
Digital LCD |