Learn More
Fisherbrand™ Traceable™ Digital Thermometer with Stainless-Steel Stem and 0.25 in. LCD Screen
Traceable™ Digital Thermometer used to measure temperature of gases, food, and semisolids.
Brand:  Fisherbrand™ 93676-96
 View more versions of this product
Product Code. 11759725
  • £118.00 / Each
Explore more special offers
Description
Description
Traceable™ Digital Thermometer used to measure temperature of gases, food, and semisolids.
  • Short-stem piercing models are ideal for carrying in a pocket
  • Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
  • Thermometers with longer sampling rates allow thermometer to be removed from sample before reading—ideal when display is difficult to read in measuring position
  • For use in cuvets, soil, incubators, petroleum products and foods
  • Traceable to standards provided by NIST
  • Waterproof thermometers (see Models 14-648-43, -44) are ideal for wet labs, wash-down areas and field use
  • 0.375 in. LCD screen thermometers available (see Cat. No.14-648-47); also available with protective guards (see Cat. Nos. 06-664-32 and 06-664-30)
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Specifications
Specifications
ISO 17025, A2LA, NIST
2.5 cm
Measuring temperature in soil, cuvettes, test tubes, food products, incubators and petroleum hydrocarbons
8 in.
20 cm
Ultra Food/Waterproof/Piercing/Drop-proof Thermometer
1.25 oz.
3.75 in.
572°F
-58°F
-58°F to +572°F
±0.4°C
9.5 cm
1 in.
0.19 in.
Battery, Stainless steel probe and Traceable certificate
240 mm
Stainless Steel
0.1°C
35 g
9.5 in.
300°C
-50°C
-50°C to +300°C
Digital LCD