Learn More
Fisherbrand™ Traceable™ Noncontact Infrared Thermometer
Traceable™ Noncontact Infrared Thermometer measures infrared surface temperatures with view fields of 6:1 to 8:1.
Brand:  Fisherbrand™ 90008-81
 View more versions of this product
Product Code. 11729785
  • £342.00 / Each
Explore more special offers
Description
Description
Traceable™ Noncontact Infrared Thermometer measures infrared surface temperatures with view fields of 6:1 to 8:1.
  • If a unit has a view field of 7:1 and the thermometer is held 14 in. (35.6 cm) from object, it reads infrared radiation emitting from a 2 in. dia. (5.1 cm) spot
  • Switch to view either Celsius or Fahrenheit scales Laser sighting beam permits aiming precisely at the target surface
  • 15-077-57 and 06-664-39 assure accurate measurements using adjustable control to account for differences in surface infrared emissivity of objects
  • One-second sampling, backlighting button, data hold, low-battery indicator and dual ranges
Note: Not meant for medical use.
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Specifications
Specifications
ISO 17025, A2LA, NIST
Two AAA Batteries, Wrist Strap
178 mm
1 sec.
6 oz.
932°F
-76°F
-76°F to +932°F
AAA
Digital LCD
Petroleum, electronics, plant and field use. (Not meant for medical use.)
7 in.
178 mm
0.17 kg
500°C
-60°C
-60°C to +500°C
±2°C or 2%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products