Learn More
Traceable™ One-Shot™ Conductivity Calibration Standards Certified Reference Material Producer (CRM), Fisherbrand™
Ideal for lab or field applications
Brand: Fisherbrand 98769-28
35.02 GBP valid until 2025-03-14
Use promo code "24142" to get your promotional price.
Description
Ideal for lab or field applications
Single-use conductivity standards calibrate all conductivity meters and probes for maximum accuracy. Eliminate concern about external container contamination. Calibration is made in the standard vial.
Calibration within container — zero contamination
- Traceable to NIST (National Institute of Standards and Technology)
- Accuracy: certificate stated uncertainty or ±0.25 microsiemen or ±0.25%
- 100% compatible with all conductivity meters
- A2LA accreditations ISO 34 and ISO 17025 plus ISO 9001
- Includes an individual temperature chart and step-by-step calibration instructions
- Units labeled (micromhos/microsiemens, ohms, parts per million/TDS)
- Single use Eliminates external contamination concerns Each standard contains 100mL
Assortment (09-328-20) contains one each of 09-328-6, -7, -8, -9, -10, and -12
Order Info
Each bottle contains 100mL
Specifications
Conductivity Standard | |
100 mL | |
Clear | |
Conductivity Meter | |
1000 μ℧ | |
Soluble in water |
6/Pk. | |
Calibration Solution | |
Conductivity Standard | |
7 | |
Calibration of Conductivity Instrument |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Traceable® Conductivity Standard
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use appropriate media to extinguish.
Supplemental information
- MIXTURE LIST-Contains : n-propanol
Special Offers
-
Multiple Manufacturers
Incredible Savings on Peptide and Protein Mass Spectrometry Testing Products
Explore our new Peptide and Protein Mass Spectrometry Testing Products PromotionPromo Code: 24142Expires: 2025-03-14
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
46Hours
22Minutes
6Seconds
27
Your input is important to us. Please complete this form to provide feedback related to the content on this product.