missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ One-Shot™ Conductivity Calibration Standards Certified Reference Material Producer (CRM), Fisherbrand™

Ideal for lab or field applications

Brand:  Fisherbrand 98769-29

51.85 GBP valid until 2024-12-31
Use promo code "21757" to get your promotional price.


 View more versions of this product

Product Code. 11794246

  • £72.95 / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ideal for lab or field applications

Single-use conductivity standards calibrate all conductivity meters and probes for maximum accuracy. Eliminate concern about external container contamination. Calibration is made in the standard vial.

Calibration within container — zero contamination

  • Traceable to NIST (National Institute of Standards and Technology)
  • Accuracy: certificate stated uncertainty or ±0.25 microsiemen or ±0.25%
  • 100% compatible with all conductivity meters
  • A2LA accreditations ISO 34 and ISO 17025 plus ISO 9001
  • Includes an individual temperature chart and step-by-step calibration instructions
  • Units labeled (micromhos/microsiemens, ohms, parts per million/TDS)
  • Single use Eliminates external contamination concerns Each standard contains 100mL

Assortment (09-328-20) contains one each of 09-328-6, -7, -8, -9, -10, and -12

Order Info

Each bottle contains 100mL

Specifications

Specifications

6/Pk.
100 mL
Conductivity Calibration Standard
10,000 μ℧
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.