Learn More
Fisherbrand™ TraceableGO™ Temperature Data Logger with Bluetooth™, Calibration and Bullet Probe
Easily access data from any location with Bluetooth™ data loggers.
Brand:  Fisherbrand™ 61011-60
95.39 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 16394446
  • £104.34 / Each
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Critical Information (Temperature, Trip Duration, MIN/MAX, Battery Level and Alarms) displayed on LCD at all times Simple, single button use
  • Safely transport reagents, clinical samples, vaccines, pharmaceuticals and food items to any location
  • Compatible with TraceableGO or TraceableLIVE Mobile Apps:
    • Once the trip has ended, users may export the data
    • App immediately identifies if the contents experienced conditions outside user-defined parameters
    • View if an alarm was triggered
    • Generate an instant data report
    • Device stores up to 90 alarm events, records readings once every 5 minutes by default, or the user may adjust the reading interval between one minute to once every 12 hours
    • Logging duration is 227 days (7.5 months)
  • TraceableGO™ Bluetooth devices are pre-programmed with alarm values
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    46
    Hours
    22
    Minutes
    23
    Seconds
    53
Product Content Correction
Product Title
Fisherbrand™ TraceableGO™ Temperature Data Logger with Bluetooth™, Calibration and Bullet Probe > TraceableGO™ Temperature Data Logger; Probe Type: Bullet Probe
Option to subscribe to cloud-based TraceableLIVE™ service to upload data to cloud.