Learn More
Fisherbrand™ TraceableLIVE™ Subscription Coupon Codes
TraceableLIVE™ Subscription Coupon Codes with simplified cloud monitoring for critical environments.
Brand: Fisherbrand™
Additional Details : Weight : 0.22680kg
Description
TraceableLIVE™-capable products securely monitor your critical environments and notify your smartphone, tablet, or PC should anything go wrong. Enjoy notifications for peace of mind- everyone on your team can receive e-mail, text, and push notifications on the environments you monitor.
- Remotely view data logging history
- Effortlessly generate reports, and control alarm parameters
- Traceable™ products come calibrated to NIST standards, helping you meet quality and regulatory requirements
- Digital data reports sent with ease whenever you need them
- Customize your report with location, device, date range, and report format
TraceableLIVE Subscription Cost through traceablelive.com:
- Monthly Payment Plan: $8.99 per device for 1-9 devices or $7.20 per device for 10+ devices
- Yearly Payment Plan: $100 per device for 1-9 devices or $80 per device for 10+ devices
- Demo TraceableLive software at traceablelive.com
Specifications
Compatible Traceable Devices | |
Coupon Code | |
Card with Unique Coupon Code and QR Code |
2 Year Standard Subscription Coupon Code | |
Cloud Monitoring |