Learn More
Thermo Scientific™ TraceGOLD TG-1301MS GC Columns
Provides low-bleed, excellent reproducibility and column-to-column consistency even with sensitive detectors like ECDs and MS.
£235.21 - £1178.99
Specifications
Stationary Phase | TraceGOLD™ TG-1301 MS |
---|---|
USP Type | G43 |
Max. Temperature | 260/280°C |
Polarity | Mid-Polar |
Product Code | Brand | Diameter (Metric) | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
10259834
|
Thermo Scientific™
26091-1420 |
0.25 mm | 0.25 μm | 30 m |
£538.22
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||
10025834
|
Thermo Scientific™
26091-1430 |
0.32 mm | 0.25 μm | 30 m |
£602.00
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||
10712917
|
Thermo Scientific™
26091-1540 |
0.25 mm | 0.25 μm | 60 m |
£982.36
Each |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||
10321315
|
Thermo Scientific™
26091-2960 |
0.25 mm | 1 μm | 30 m |
£548.66
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
10381355
|
Thermo Scientific™
26091-2970 |
0.32 mm | 1 μm | 30 m |
£590.41
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10117334
|
Thermo Scientific™
26091-2980 |
0.53 mm | 1 μm | 30 m |
£690.64
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
10127334
|
Thermo Scientific™
26091-3080 |
0.25 mm | 1 μm | 60 m |
£989.13
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||
10301025
|
Thermo Scientific™
26091-3330 |
0.25 mm | 1.4 μm | 60 m |
£961.07
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||
10487134
|
Thermo Scientific™
26091-3350 |
0.32 mm | 1.5 μm | 30 m |
£597.87
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||
10229134
|
Thermo Scientific™
26091-3390 |
0.32 mm | 1.8 μm | 30 m |
£590.41
Each |
In Stock
Log in to see stock. |
|||
10784637
|
Thermo Scientific™
26091-3410 |
0.32 mm | 1.8 μm | 60 m |
£1073.34
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
10406954
|
Thermo Scientific™
26091-3960 |
0.53 mm | 3 μm | 30 m |
£658.01
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||
10565815
|
Thermo Scientific™
26091-4080 |
0.53 mm | 3 μm | 60 m |
£1069.83
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||
10229184
|
Thermo Scientific™
26091-2250 |
0.53 mm | 0.5 μm | 30 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10036124
|
Thermo Scientific™
26091-2350 |
0.25 mm | 0.5 μm | 60 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10792147
|
Thermo Scientific™
26091-2360 |
0.32 mm | 0.5 μm | 60 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10391015
|
Thermo Scientific™
26091-2370 |
0.53 mm | 0.5 μm | 60 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10320835
|
Thermo Scientific™
26091-2840 |
0.25 mm | 1 μm | 15 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10075984
|
Thermo Scientific™
26091-2850 |
0.32 mm | 1 μm | 15 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10036034
|
Thermo Scientific™
26091-2860 |
0.53 mm | 1 μm | 15 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10187544
|
Thermo Scientific™
26091-1310 |
0.32 mm | 0.25 μm | 15 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10219134
|
Thermo Scientific™
26091-1440 |
0.53 mm | 0.25 μm | 30 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10514465
|
Thermo Scientific™
26091-1300 |
0.25 mm | 0.25 μm | 15 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10660056
|
Thermo Scientific™
26091-1550 |
0.32 mm | 0.25 μm | 60 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10230295
|
Thermo Scientific™
26091-3090 |
0.32 mm | 1 μm | 60 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10641026
|
Thermo Scientific™
26091-3100 |
0.53 mm | 1 μm | 60 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10774637
|
Thermo Scientific™
26091-1560 |
0.53 mm | 0.25 μm | 60 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10792337
|
Thermo Scientific™
26091-3340 |
0.53 mm | 1.5 μm | 15 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10167894
|
Thermo Scientific™
26091-2110 |
0.25 mm | 0.5 μm | 15 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10487324
|
Thermo Scientific™
26091-3360 |
0.53 mm | 1.5 μm | 30 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10046314
|
Thermo Scientific™
26091-2120 |
0.32 mm | 0.5 μm | 15 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10499244
|
Thermo Scientific™
26091-2130 |
0.53 mm | 0.5 μm | 15 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10341305
|
Thermo Scientific™
26091-2230 |
0.25 mm | 0.5 μm | 30 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10076074
|
Thermo Scientific™
26091-2240 |
0.32 mm | 0.5 μm | 30 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10437724
|
Thermo Scientific™
26091-4090 |
0.53 mm | 3 μm | 105 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10117424
|
Thermo Scientific™
26091-4900 |
0.53 mm | 3 μm | 75 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Thermo Scientific™ TraceGOLD TG-1301MS GC Columns provide low-bleed, excellent reproducibility and column-to-column consistency even with sensitive detectors like ECDs and MS.
- Low to mid-polarity phase, 6% cyanopropylphenyl/94% dimethyl polysiloxane
- Long lifetime
- Excellent inertness
- Equivalent to USP G43 phase
- Similar to: Rtx-1301, DB-1301, BP-624, HP-1301, HP-624, SPB-1301, SPB-624, VP-1301, BF-624ms, CP-1301, CP-Select 624 CB
Recommended for:
- Alcohols
- Volatile organics
- Oxygenates
- Residual solvents
Specifications
TraceGOLD™ TG-1301 MS | |
G43 | |
260/280°C | |
Mid-Polar |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.