Learn More
Thermo Scientific™ TraceGOLD TG-VRX GC Columns
Thermo Scientific™ TraceGOLD TG-VRX GC Columns provide good resolution and fast analysis times for volatile compounds.
Brand:  Thermo Scientific™ 26081-4090
 View more versions of this product
Product Code. 11571164
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ TraceGOLD TG-VRX GC Columns provide good resolution and fast analysis times for volatile compounds.
  • Proprietary stationary phase
  • Optimized dimensions for analysis of volatile organic pollutants
  • Ideally suited for US EPA Methods 502.2, 524.2, 8021, and 8260
  • Similar to DB-VRX, Rtx-VRX
Recommended for:
  • Volatile organic pollutants
  • Aromatics
TRUSTED_SUSTAINABILITY
Specifications
Specifications
260°C
0.53 mm
GC/MS for Volatile Organic Pollutants, GC/MS for US EPA Method 8021 Compounds
105 m
105m x 0.53mm x 3.00um
0.53 mm
TraceGOLD™ TG-VRX
3 μm
Non-Polar
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ TraceGOLD TG-VRX GC Columns > 105m x 0.53mm x 3.00μm