missing translation for 'onlineSavingsMsg'
Learn More
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid Monohydrate, 98%, Thermo Scientific Chemicals Product Code.: 10782054

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid Monohydrate, 98%, Thermo Scientific Chemicals

Product Code. 10782054
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 10782054

Brand: Thermo Scientific Acros 155005000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

trans-1,2-Diaminocyclohexane-N,N,N,N-tetraacetic acid monohydrate, 98%, C14H24N2O9, CAS Number-125572-95-4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125572-95-4
Molecular Formula C14H20N2O8
Molecular Weight (g/mol) 344.32
MDL Number MFCD00149243,MFCD00066429,MFCD00003845
InChI Key FCKYPQBAHLOOJQ-NXEZZACHSA-L
Synonym 2,2',2,2'-trans-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, dcyta, trans-1,2-diaminocyclohexane-n,n,n',n'-tetraacetic acid monohydrate, 1,2-cyclohexanedinitrilotetraacetic acid, 2,2',2,2'-1r,2r-rel-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, trans-1,2-cyclohexanediaminetetraacetic acid monohydrate, cdta monohydrate, ctda monohydrate, chel-cd, chel r-cd
PubChem CID 2723844
SMILES [O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O

Specifications

Melting Point >210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Complexometry)
Packaging Plastic bottle
Linear Formula C6H10[N(CH2CO2H)2]2·H2O
Quantity 500 g
Beilstein 13,III,1
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in most common organic solvents, soluble in alkali solutions
IUPAC Name 2-[[(1R,2R)-2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate
Formula Weight 364.35
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid Monohydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.