Learn More
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate, ACS, 97.5%-100.5%, Thermo Scientific Chemicals
Description
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate is an analytical reagent and a complexating agent for sodium. It acts as a ligand to prepare lanthanide shift reagents. It is involved in the separation and determination of iron(III)-dimethyldithiocarbamate, manganese(II)-ethylenebisdithiocarbamate and zinc(II)-ethylenebisdithiocarbamate by sensitive capillary electrophoretic ultraviolet method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 213°C to 216°C |
Assay Percent Range | 97.5 to 100.5% |
Quantity | 25 g |
Beilstein | 3222748 |
Solubility Information | Slightly soluble in water. Soluble in 1N sodium hydroxide and alkali solutions. Insoluble in most common organic solvents. |
Formula Weight | 364.35 (346.34 Anhydrous) |
Percent Purity | 97.5 to 100.5% |
Grade | ACS |
Physical Form | Solid |
Chemical Name or Material | trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate, For analysis ACS |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N06L009 | Certificate of Analysis | 10/02/2025 | 036697.06,036697.14,036697.36,036697.22 |
Y21K005 | Certificate of Analysis | 26/11/2024 | 036697.06,036697.14,036697.36,036697.22 |
T24K037 | Certificate of Analysis | 01/08/2024 | 036697.06,036697.14,036697.36,036697.22 |
S10K013 | Certificate of Analysis | 20/06/2024 | 036697.06,036697.14,036697.36,036697.22 |
Q08K011 | Certificate of Analysis | 17/04/2024 | 036697.06,036697.14,036697.36,036697.22 |
Safety and Handling

- trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P362-Take off contaminated clothing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.