missing translation for 'onlineSavingsMsg'
Learn More

trans-1,3-Dimethylcyclopentane, SPEX CertiPrep™

Product Code. 16300238 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
trans-1,3-Dimethylcyclopentane1759-58-61000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 1759-58-6, 67-56-1
Packaging 2mL amber prescored glass ampule
CAS Min % 0.1, 99.9
Quantity 1 mL
CAS Max % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Molecular Formula C7H14
InChI Key XAZKFISIRYLAEE-RNFRBKRXSA-N
SMILES CC1CCC(C1)C
IUPAC Name (1R,3R)-1,3-dimethylcyclopentane
Molecular Weight (g/mol) 98.189
PubChem CID 15656
Solvent or Matrix Methanol-P&T
Chemical Name or Material trans-1,3-Dimethylcyclopentane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • trans-1,3-Dimethylcyclopentane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methanol

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.