missing translation for 'onlineSavingsMsg'
Learn More

trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane, 97%, Thermo Scientific Chemicals

£190.95 - £540.69

Chemical Identifiers

CAS 1212405-03-2
Molecular Formula C12H22BrNO2
Molecular Weight (g/mol) 292.217
MDL Number MFCD12022626
InChI Key XJDUGNDNCQMMBK-NXEZZACHSA-N
Synonym tert-butyl trans-3-bromomethyl-cyclohexylcarbamate, t-butyl trans-3-bromomethyl cyclohexyl carbamate, tert-butyl n-1r,3r-3-bromomethyl cyclohexyl carbamate
PubChem CID 46172881
IUPAC Name tert-butyl N-[(1R,3R)-3-(bromomethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)CBr
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15487706
SDS View Documents
Thermo Scientific Alfa Aesar
H62670.MD
250 mg
£190.95
250mg
Please to purchase this item. Need a web account? Register with us today!
15477706
SDS View Documents
Thermo Scientific Alfa Aesar
H62670.03
1 g
£540.69
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.