missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1-(Boc-amino)-4-(4-methyl-1-piperazinyl)cyclohexane, 97%, Thermo Scientific™
£111.17 - £333.52
Chemical Identifiers
CAS | 1119283-74-7 |
---|---|
Molecular Formula | C16H31N3O2 |
Molecular Weight (g/mol) | 297.443 |
InChI Key | HVPDOBVAGJYPNN-UHFFFAOYSA-N |
Synonym | tert-butyl trans-4-4-methylpiperazin-1-yl cyclohexylcarbamate, tert-butyl n-1r,4r-4-4-methylpiperazin-1-yl cyclohexyl carbamate |
PubChem CID | 58236010 |
IUPAC Name | tert-butyl N-[4-(4-methylpiperazin-1-yl)cyclohexyl]carbamate |
SMILES | CC(C)(C)OC(=O)NC1CCC(CC1)N2CCN(CC2)C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1119283-74-7 | |
297.443 | |
tert-butyl trans-4-4-methylpiperazin-1-yl cyclohexylcarbamate, tert-butyl n-1r,4r-4-4-methylpiperazin-1-yl cyclohexyl carbamate | |
tert-butyl N-[4-(4-methylpiperazin-1-yl)cyclohexyl]carbamate |
C16H31N3O2 | |
HVPDOBVAGJYPNN-UHFFFAOYSA-N | |
58236010 | |
CC(C)(C)OC(=O)NC1CCC(CC1)N2CCN(CC2)C |
Specifications
1119283-74-7 | |
tert-butyl trans-4-4-methylpiperazin-1-yl cyclohexylcarbamate, tert-butyl n-1r,4r-4-4-methylpiperazin-1-yl cyclohexyl carbamate | |
CC(C)(C)OC(=O)NC1CCC(CC1)N2CCN(CC2)C | |
297.443 | |
297.43 | |
Solid |
C16H31N3O2 | |
HVPDOBVAGJYPNN-UHFFFAOYSA-N | |
tert-butyl N-[4-(4-methylpiperazin-1-yl)cyclohexyl]carbamate | |
58236010 | |
97% | |
trans-1-(Boc-amino)-4-(4-methyl-1-piperazinyl)cyclohexane |