missing translation for 'onlineSavingsMsg'
Learn More

trans-2,3-Dimethylacrylic Acid, 98%

Product Code. 10308753 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10308753

Brand: Thermo Scientific Acros 138975000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-59-1 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-59-1
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00066864
InChI Key UIERETOOQGIECD-ONEGZZNKSA-N
Synonym tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid
PubChem CID 125468
ChEBI CHEBI:9592
IUPAC Name (E)-2-methylbut-2-enoic acid
SMILES C\C=C(/C)C(O)=O

Specifications

Melting Point 61.0°C to 65.0°C
Color White to Beige
Density 0.9600g/mL
Boiling Point 198.4°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3CH=C(CH3)CO2H
Quantity 500 g
Beilstein 02, 430
Merck Index 15, 9589
Specific Gravity 0.96
Solubility Information Solubility in water: soluble in hot water. Other solubilities: 1g/10mL methanol,soluble in alcohol and ether
Formula Weight 100.12
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material trans-2, 3-Dimethylacrylic acid
Show More Show Less
compliance-icons
Product Identifier
  • trans-2,3-Dimethylacrylic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.