missing translation for 'onlineSavingsMsg'
Learn More

trans-2,3-Epoxybutane, 97%, Thermo Scientific Chemicals

Product Code. 11349749
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

A higher order cuprate reacted with trans-2,3-epoxybutane a high yield both counted on the epoxide (96%) and on the haloalkane (85%). Enolate derived from trans-2,3-Epoxybutan has chiral recognition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A higher order cuprate reacted with trans-2,3-epoxybutane a high yield both counted on the epoxide (96%) and on the haloalkane (85%). Enolate derived from trans-2,3-Epoxybutan has chiral recognition.

Solubility
Soluble in water(95g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21490-63-1
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00005129
InChI Key PQXKWPLDPFFDJP-QWWZWVQMSA-N
Synonym trans-2,3-dimethyloxirane, trans-2,3-epoxybutane, 2r,3r-2,3-dimethyloxirane, oxirane, 2,3-dimethyl-, trans, trans-2-butene oxide, butane, 2,3-epoxy-, trans, e-2r,3r-epoxybutane, trans-, a-butylene oxide, trans-2r,3r-epoxybutane, 2,3-dimethyloxirane, trans
PubChem CID 6432237
IUPAC Name (2R,3R)-2,3-dimethyloxirane
SMILES CC1C(O1)C

Specifications

Density 0.804
Boiling Point 54°C to 55°C
Flash Point −26°C (−14°F)
Odor Recognizable
Refractive Index 1.373
Quantity 25 g
UN Number UN3271
Beilstein 79772
Sensitivity Moisture sensitive
Solubility Information Soluble in water(95g/L).
Formula Weight 72.11
Percent Purity 97%
Chemical Name or Material trans-2,3-Epoxybutane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'