missing translation for 'onlineSavingsMsg'
Learn More

trans-2,4-Dichlorocinnamic acid, 98%, Thermo Scientific™

Product Code. 11444914
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20595-45-3
Molecular Formula C9H6Cl2O2
Molecular Weight (g/mol) 217.045
MDL Number MFCD00004373
InChI Key MEBWABJHRAYGFW-DUXPYHPUSA-N
Synonym 2,4-dichlorocinnamic acid, trans-2,4-dichlorocinnamic acid, e-3-2,4-dichlorophenyl acrylic acid, cinnamic acid, 2,4-dichloro, 3-2,4-dichlorophenyl-2-propenoic acid, unii-c09kk5d2t2, 2-propenoic acid, 3-2,4-dichlorophenyl-, 2e, 2e-3-2,4-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,4-dichlorophenyl, 2e-3-2,4-dichlorophenyl prop-2-enoic acid
PubChem CID 688026
IUPAC Name (E)-3-(2,4-dichlorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1Cl)Cl)C=CC(=O)O

Specifications

Melting Point 233°C to 235°C
Quantity 25 g
Beilstein 1369152
Formula Weight 217.05
Percent Purity 98%
Chemical Name or Material trans-2,4-Dichlorocinnamic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'