missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Aminomethyl-1-cyclohexanol, 99+%, Thermo Scientific Chemicals

Product Code. 10359340
Click to view available options
Quantity:
1 g
2.5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5691-09-8
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.2
MDL Number MFCD06213163
InChI Key JGKFBZBVCAWDFD-NKWVEPMBSA-O
Synonym trans-2-aminomethyl-1-cyclohexanol, trans-2-aminomethyl-cyclohexanol, 1r,2s-2-aminomethyl cyclohexan-1-ol, jgkfbzbvcawdfd-nkwvepmbsa-n, 1r,2s-2-aminomethyl cyclohexanol, 1r,2s---trans-2-aminomethyl cyclohexanol, trans-2-aminomethyl cyclohexanol, 2-aminomethyl cyclohexanol, e, cyclohexanol, 2-aminomethyl-, trans, 1r-2alpha-aminomethylcyclohexan-1beta-ol
PubChem CID 2724658

Specifications

Color Colorless
Boiling Point 226°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula OHC6H10CH2NH2
Refractive Index 1.4905 to 1.4925
Quantity 2.5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,ethanol,methanol
Formula Weight 129.2
Percent Purity 99+%
Physical Form Liquid After Melting
Chemical Name or Material trans-2-Aminomethyl-1-cyclohexanol, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • trans-2-Aminomethyl-1-cyclohexanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'