missing translation for 'onlineSavingsMsg'
Learn More
trans-2-Hexene, 99%, Thermo Scientific Chemicals Product Code.: 11495956

trans-2-Hexene, 99%, Thermo Scientific Chemicals

Product Code. 11495956
2.5 g
Click to view available options
Quantity:
2.5 g
10 g
This item is not returnable. View return policy

Product Code. 11495956

Brand: Thermo Scientific Alfa Aesar B22289.88

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

trans-2-Hexene is used as a co monomer in the production of polyethylene. The use of trans-2-hexene, which is derived from 3-hexyne using sodium in liquid ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-2-Hexene is used as a co monomer in the production of polyethylene. The use of trans-2-hexene, which is derived from 3-hexyne using sodium in liquid ammonia.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4050-45-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00009473
InChI Key RYPKRALMXUUNKS-HWKANZROSA-N
Synonym trans-2-hexene, e-2-hexene, 2-hexene, e, hexene, isomer, hexene, isomers, unii-2tz30ggg1a, 2e-hexene, e-hex-2-ene, 2tz30ggg1a, 2-hexene, 2e
PubChem CID 639661
SMILES CCC\C=C\C

Specifications

Melting Point -99°C
Density 0.674
Boiling Point 68°C to 69°C
Flash Point −20°C (−4°F)
Odor Mild
Refractive Index 1.394
Quantity 2.5 g
UN Number UN3295
Beilstein 1719020
Solubility Information Difficult to mix.
IUPAC Name (2E)-hex-2-ene
Formula Weight 84.16
Percent Purity 99%
Chemical Name or Material trans-2-Hexene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-2-Hexene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.