Learn More
trans-2-Methyl-2-pentenoic acid, 97%, Thermo Scientific™
£15.70 - £45.76
Chemical Identifiers
CAS | 16957-70-3 |
---|---|
Molecular Formula | C6H10O2 |
Molecular Weight (g/mol) | 114.144 |
MDL Number | MFCD00002655 |
InChI Key | JJYWRQLLQAKNAD-SNAWJCMRSA-N |
Synonym | 2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid |
PubChem CID | 5365909 |
IUPAC Name | (E)-2-methylpent-2-enoic acid |
SMILES | CCC=C(C)C(=O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
16957-70-3 | |
114.144 | |
JJYWRQLLQAKNAD-SNAWJCMRSA-N | |
5365909 | |
CCC=C(C)C(=O)O |
C6H10O2 | |
MFCD00002655 | |
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid | |
(E)-2-methylpent-2-enoic acid |
Specifications
16957-70-3 | |
0.979 | |
108°C (226°F) | |
C2H5CH=C(CH3)CO2H | |
MFCD00002655 | |
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid | |
CCC=C(C)C(=O)O | |
114.144 | |
114.14 | |
trans-2-Methyl-2-pentenoic acid |
26°C to 28°C | |
123°C to 125°C (30 mmHg) | |
C6H10O2 | |
1.46 | |
UN3261 | |
JJYWRQLLQAKNAD-SNAWJCMRSA-N | |
(E)-2-methylpent-2-enoic acid | |
5365909 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 241-026-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures