missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Octenal, 95%

Product Code. 11423108
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11423108

Brand: Thermo Scientific Alfa Aesar A18830.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2548-87-0 | C8H14O | 126.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Used in flavoring tangerine, mandarin, cherry, melon and orange and also as a fragrances agent. It causes defects for locomotion in green fluorescent protein (GFP). It is an antimicrobial agent that destroys fungi by suppressing their ability to grow or reproduce.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol and fixed oils.

Notes
Air Sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2548-87-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00007011
InChI Key LVBXEMGDVWVTGY-VOTSOKGWSA-N
Synonym trans-2-octenal, e-oct-2-enal, e-2-octenal, 2-octenal, trans-2-octen-1-al, e-2-octen-1-al, 2-octen-1-al, 2-octenel, 2-octenal, e, 2-octenal, 2e
PubChem CID 5283324
ChEBI CHEBI:61748
IUPAC Name (E)-oct-2-enal
SMILES CCCCCC=CC=O

Specifications

Density 0.84
Boiling Point 84°C to 86°C (19 mmHg)
Flash Point 71°C (160°F)
Odor Pleasant
Refractive Index 1.45
Quantity 25 g
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol and fixed oils.
Formula Weight 126.2
Percent Purity 95%
Chemical Name or Material trans-2-Octenal
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.