missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-3,4,5-Trifluoro-4'-(4-n-propylcyclohexyl)biphenyl, 99%, Thermo Scientific™
£47.60 - £178.00
Chemical Identifiers
CAS | 132123-39-8 |
---|---|
Molecular Formula | C21H23F3 |
Molecular Weight (g/mol) | 332.41 |
MDL Number | MFCD09839001 |
InChI Key | RRKRBRVTKIRLHH-UHFFFAOYSA-N |
Synonym | 3,4,5-trifluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 4'-trans-4-propylcyclohexyl-3,4,5-trifluorobiphenyl, 1,2,3-trifluoro-5-4-4-propylcyclohexyl phenyl benzene, 4'-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, 3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, trans-3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, 4-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, trans-3,4,5-trifluoro-4'-4-n-propylcyclohexyl biphenyl, 1,1'-biphenyl, 3,4,5-trifluoro-4'-trans-4-propylcyclohexyl |
PubChem CID | 15014921 |
IUPAC Name | 1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)phenyl]benzene |
SMILES | CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C(=C3)F)F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is employed as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
132123-39-8 | |
332.41 | |
RRKRBRVTKIRLHH-UHFFFAOYSA-N | |
15014921 | |
CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C(=C3)F)F)F |
C21H23F3 | |
MFCD09839001 | |
3,4,5-trifluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 4'-trans-4-propylcyclohexyl-3,4,5-trifluorobiphenyl, 1,2,3-trifluoro-5-4-4-propylcyclohexyl phenyl benzene, 4'-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, 3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, trans-3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, 4-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, trans-3,4,5-trifluoro-4'-4-n-propylcyclohexyl biphenyl, 1,1'-biphenyl, 3,4,5-trifluoro-4'-trans-4-propylcyclohexyl | |
1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)phenyl]benzene |
Specifications
132123-39-8 | |
MFCD09839001 | |
Slightly soluble in water. | |
CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C(=C3)F)F)F | |
332.41 | |
332.4 | |
trans-3,4,5-Trifluoro-4'-(4-n-propylcyclohexyl)biphenyl |
C21H23F3 | |
3,4,5-trifluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 4'-trans-4-propylcyclohexyl-3,4,5-trifluorobiphenyl, 1,2,3-trifluoro-5-4-4-propylcyclohexyl phenyl benzene, 4'-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, 3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, trans-3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, 4-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, trans-3,4,5-trifluoro-4'-4-n-propylcyclohexyl biphenyl, 1,1'-biphenyl, 3,4,5-trifluoro-4'-trans-4-propylcyclohexyl | |
RRKRBRVTKIRLHH-UHFFFAOYSA-N | |
1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)phenyl]benzene | |
15014921 | |
99% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures