missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(Boc-amino)cyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15447976
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939400-34-7
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD06657680
InChI Key KLCYDBAYYYVNFM-UHFFFAOYSA-N
Synonym cis-3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, trans-3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, 3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, trans-3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, 3-boc-amino cyclobutanecarboxylic acid, 3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, cis-3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, cis-3-boc-amino cyclobutanecarboxylic acid, 3-tert-butoxy carbonyl amino cyclobutane-1-carboxylic acid, cis-3-boc-aminocyclobutanecarboxylic acid
PubChem CID 24720963
IUPAC Name 3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CC(C1)C(=O)O

Specifications

Formula Weight 215.24
Percent Purity 97%
Quantity 250 mg
Chemical Name or Material trans-3-(Boc-amino)cyclobutanecarboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'