missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-3-(Boc-amino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical and chemical research.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical and chemical research.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
IUPAC Name | 3-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylic acid |
Formula Weight | 243.29 |
Percent Purity | 97% |
Quantity | 1 g |
Chemical Name or Material | trans-3-(Boc-amino)cyclohexanecarboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'