Learn More
trans-3-chloroacrylic acid, 99%, Thermo Scientific Chemicals
£58.30 - £216.00
Chemical Identifiers
CAS | 2345-61-1 |
---|---|
Molecular Formula | C3H3ClO2 |
Molecular Weight (g/mol) | 106.51 |
MDL Number | MFCD00064237 |
InChI Key | MHMUCYJKZUZMNJ-OWOJBTEDSA-N |
Synonym | trans-3-chloroacrylic acid, e-3-chloroacrylic acid, 3-chloroacrylic acid, 3-chloroprop-2-enoic acid, acrylic acid, 3-chloro-, trans, acrylic acid, 3-chloro-, e, trans-3-chloropropenoic acid, trans-beta-chloroacrylic acid, 2-propenoic acid, 3-chloro-, e, ccris 3546 |
PubChem CID | 638124 |
IUPAC Name | (E)-3-chloroprop-2-enoic acid |
SMILES | C(=CCl)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10774021
|
Thermo Scientific Acros
108550010 |
1 g | Glass bottle |
£58.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10197793
|
Thermo Scientific Acros
108550050 |
5 g | Glass bottle |
£216.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2345-61-1 | |
106.51 | |
MHMUCYJKZUZMNJ-OWOJBTEDSA-N | |
638124 | |
C(=CCl)C(=O)O |
C3H3ClO2 | |
MFCD00064237 | |
trans-3-chloroacrylic acid, e-3-chloroacrylic acid, 3-chloroacrylic acid, 3-chloroprop-2-enoic acid, acrylic acid, 3-chloro-, trans, acrylic acid, 3-chloro-, e, trans-3-chloropropenoic acid, trans-beta-chloroacrylic acid, 2-propenoic acid, 3-chloro-, e, ccris 3546 | |
(E)-3-chloroprop-2-enoic acid |
Specifications
2345-61-1 | |
Beige-Gray to Gray | |
99% | |
ClCH=CHCO2H | |
02, 400 | |
MHMUCYJKZUZMNJ-OWOJBTEDSA-N | |
(E)-3-chloroprop-2-enoic acid | |
638124 | |
99% | |
trans-3-chloroacrylic acid |
82°C to 85°C | |
Authentic | |
C3H3ClO2 | |
MFCD00064237 | |
trans-3-chloroacrylic acid, e-3-chloroacrylic acid, 3-chloroacrylic acid, 3-chloroprop-2-enoic acid, acrylic acid, 3-chloro-, trans, acrylic acid, 3-chloro-, e, trans-3-chloropropenoic acid, trans-beta-chloroacrylic acid, 2-propenoic acid, 3-chloro-, e, ccris 3546 | |
C(=CCl)C(=O)O | |
106.51 | |
106.51 | |
Powder or Flakes |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 219-070-
- trans-3-Chloroacrylic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only